Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human EpCAM / TROP1 Protein, His Tag & Avi Tag (Avitag™);HEK293;200UG;EPM,EPCAM,DIAR5,DIAR-5,EGP-2,EGP2,EGP314,EGP-314,EGP40,EGP-40,ESA,GA733-2,HNPCC8,HNPCC-8,KS1/4,KSA,M4S1,MIC18,MIC-18,MK-1,MK1,TACSTD1,TACSTD-1,TROP1,TROP-1,CD326,CD-326
Catalog No. 501658507
Encompass_Preferred
Recombinant Protein;Biotinylated Human EpCAM / TROP1 Protein, His Tag & Avi Tag (Avitag™);200UG;Biotinylated Human EpCAM, His Tag (EPM-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 30 kDa. The protein migrates as 36-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8507 Supplier ACROBiosystems Supplier No. EPMH82E8200UG
Recombinant Protein;Biotinylated Human EpCAM / TROP1 Protein, His Tag & Avi Tag (Avitag™);200UG;Biotinylated Human EpCAM, His Tag (EPM-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 30 kDa. The protein migrates as 36-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.