Learn More
Cayman Chemical EPCRCD201 RabbIt MoncLNal 50uL
SDP
Catalog No. 502917419
Encompass_Preferred
An inhibitor of RNA polymerase I-mediated transcription binds to G-quadruplex DNA and inhibits the interaction between ribosomal DNA G-quadruplexes and nucleolin induces nucleolin mislocalization from the nucleoli to the nucleoplasm in A549 cells at 10 UM induces apoptosis in A549 and HL-60 cells at 5 UM reduces tumor growth in MDA-MB-231 and MiaPaCa-2 mouse xenograft models at 5 mg/kg
Catalog No. 50-291-7419 Supplier Cayman Chemical Supplier No. 3808050 UL
An inhibitor of RNA polymerase I-mediated transcription binds to G-quadruplex DNA and inhibits the interaction between ribosomal DNA G-quadruplexes and nucleolin induces nucleolin mislocalization from the nucleoli to the nucleoplasm in A549 cells at 10 UM induces apoptosis in A549 and HL-60 cells at 5 UM reduces tumor growth in MDA-MB-231 and MiaPaCa-2 mouse xenograft models at 5 mg/kg