Learn More
McMaster-Carr High-Strength Weather-Resistant EPDM Rubber Sheet with Crisscross Texture, 12" x 12", 1/16" Thick
Catalog No. NC2755078
Encompass
Color: BlackTemperature Range: -40° to 240° FTensile Strength: 1,500 psiFor Use Outdoors: YesCrisscross Texture
Catalog No. NC2755078 Supplier McMaster-Carr Supplier No. 3727T22
Color: BlackTemperature Range: -40° to 240° FTensile Strength: 1,500 psiFor Use Outdoors: YesCrisscross Texture