Learn More
Medchemexpress LLC HY-B1901 10mg Medchemexpress, Eperisone (Hydrochloride) CAS:56839-43-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB190110MG
Encompass_Preferred
Medchemexpress, HY-B1901 10mg Eperisone (Hydrochloride) CAS:56839-43-1 Eperisone Hydrochloride ((±)-Eperisone hydrochloride) is an antispastic agent used for treatment of diseases characterized by muscle stiffness and pain. It works by relaxing both skeletal muscles and vascularsmooth muscles, thus demonstrating avariety of effects such as reduction ofmyotonia, improvement of circulationand suppression of the pain reflex. Eperisone Hydrochloride ((±)-Eperisone hydrochloride) is a centrally acting muscle relaxant inhibiting the pain reflex pathway, having a vasodilator effect [1] [2 [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3034