Learn More
Medchemexpress LLC Eperisone (hydrochloride) | 56839-43-1 | 99.95% | 1 G
SDP
Catalog No. 5000411313
Encompass_Preferred
Eperisone hydrochloride is an orally active antispastic agent with a vasodilator effect. It is used for the research of muscle stiffness and pain. This compound is a potent and selective P2X7 receptor antagonist, also exhibiting antagonism for human P2X3. It works by relaxing both skeletal muscles and vascular smooth muscles, demonstrating several beneficial effects.
  • Reduces myotonia.
  • Improves circulation.
  • Suppresses the pain reflex.
  • Preferentially suppresses lung fibroblast activity.
  • Suppresses bleomycin-dependent pulmonary fibrosis.
  • Not toxic to alveolar epithelial cells in vitro.
Catalog No. 50-004-11313 Supplier Medchemexpress LLC Supplier No. HYB19011G
Eperisone hydrochloride is an orally active antispastic agent with a vasodilator effect. It is used for the research of muscle stiffness and pain. This compound is a potent and selective P2X7 receptor antagonist, also exhibiting antagonism for human P2X3. It works by relaxing both skeletal muscles and vascular smooth muscles, demonstrating several beneficial effects.
  • Reduces myotonia.
  • Improves circulation.
  • Suppresses the pain reflex.
  • Preferentially suppresses lung fibroblast activity.
  • Suppresses bleomycin-dependent pulmonary fibrosis.
  • Not toxic to alveolar epithelial cells in vitro.