Learn More
Medchemexpress LLC Eperisone (hydrochloride) | 56839-43-1 | 99.95% | 25 MG
SDP
Catalog No. 5000412550
Encompass_Preferred
Eperisone Hydrochloride is an orally active antispastic agent that also exhibits a vasodilator effect. It is used in research for muscle stiffness and pain. This compound is a potent and selective P2X7 receptor antagonist and also shows antagonism for human P2X3. Eperisone Hydrochloride functions by relaxing both skeletal and vascular smooth muscles, leading to various effects such as reduced myotonia, improved circulation, and suppression of the pain reflex.
  • Antispastic agent
  • Vasodilator effect
  • P2X7 receptor antagonist
  • P2X3 antagonist
  • Muscle relaxation
  • Therapeutic effects
  • Research use only
Catalog No. 50-004-12550 Supplier Medchemexpress LLC Supplier No. 25MGHYB1901
Eperisone Hydrochloride is an orally active antispastic agent that also exhibits a vasodilator effect. It is used in research for muscle stiffness and pain. This compound is a potent and selective P2X7 receptor antagonist and also shows antagonism for human P2X3. Eperisone Hydrochloride functions by relaxing both skeletal and vascular smooth muscles, leading to various effects such as reduced myotonia, improved circulation, and suppression of the pain reflex.
  • Antispastic agent
  • Vasodilator effect
  • P2X7 receptor antagonist
  • P2X3 antagonist
  • Muscle relaxation
  • Therapeutic effects
  • Research use only