Learn More
Medchemexpress LLC Eperisone hydrochloride | 56839-43-1 | MFCD00941459 | 100.0% | 500 MG
SDP
Catalog No. 5000411264
Encompass_Preferred
Eperisone Hydrochloride ((±)-Eperisone hydrochloride) is an orally active antispastic agent with a vasodilator effect, used for the research of muscle stiffness and pain. It is a potent and selectively P2X7 receptor antagonist, and also shows antagonism for human P2X3. Eperisone Hydrochloride works by relaxing both skeletal muscles and vascular smooth muscles, demonstrating a variety of effects such as reduction of myotonia, improvement of circulation and suppression of the pain reflex.
  • Orally active antispastic agent
  • Vasodilator effect
  • Used for research of muscle stiffness and pain
  • Potent and selective P2X7 receptor antagonist
  • Shows antagonism for human P2X3
  • Relaxes both skeletal muscles and vascular smooth muscles
  • Reduces myotonia
  • Improves circulation
  • Suppresses the pain reflex
Catalog No. 50-004-11264 Supplier Medchemexpress LLC Supplier No. HYB1901500MG
Eperisone Hydrochloride ((±)-Eperisone hydrochloride) is an orally active antispastic agent with a vasodilator effect, used for the research of muscle stiffness and pain. It is a potent and selectively P2X7 receptor antagonist, and also shows antagonism for human P2X3. Eperisone Hydrochloride works by relaxing both skeletal muscles and vascular smooth muscles, demonstrating a variety of effects such as reduction of myotonia, improvement of circulation and suppression of the pain reflex.
  • Orally active antispastic agent
  • Vasodilator effect
  • Used for research of muscle stiffness and pain
  • Potent and selective P2X7 receptor antagonist
  • Shows antagonism for human P2X3
  • Relaxes both skeletal muscles and vascular smooth muscles
  • Reduces myotonia
  • Improves circulation
  • Suppresses the pain reflex