Learn More
Medchemexpress LLC HY-107367A 10mg Medchemexpress, Epertinib (hydrochloride) CAS:2071195-74-7 Purity:>98%
SDP
Catalog No. 501968389
Encompass_Preferred
Medchemexpress, HY-107367A 10mg Epertinib (HCl) CAS:2071195-74-7 Epertinib hydrochloride is a potent, orally active, reversible, and selective tyrosine kinase inhibitor of EGFR, HER2 and HER4, with IC50s of 1.48 nM, 7.15 nM and 2.49 nM, respectively. Epertinib shows potent antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8389 Supplier Medchemexpress LLC Supplier No. HY107367A10MG
Medchemexpress, HY-107367A 10mg Epertinib (HCl) CAS:2071195-74-7 Epertinib hydrochloride is a potent, orally active, reversible, and selective tyrosine kinase inhibitor of EGFR, HER2 and HER4, with IC50s of 1.48 nM, 7.15 nM and 2.49 nM, respectively. Epertinib shows potent antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.