Learn More
Selleck Chemical LLC Epertinib hydrochloride
SDP
Catalog No. 5000002772
Encompass_Preferred
Epertinib hydrochloride (S-222611 hydrochloride) is a potent orally active reversible and selective tyrosine kinase inhibitor of EGFR HER2 and HER4 with IC50 of 1 48 nM 7 15 nM and 2 49 nM respectively Epertinib hydrochloride (S-222611 hydrochloride) exhibits antitumor activity
Catalog No. 50-000-02772 Supplier Selleck Chemical LLC Supplier No. S68975MG
Epertinib hydrochloride (S-222611 hydrochloride) is a potent orally active reversible and selective tyrosine kinase inhibitor of EGFR HER2 and HER4 with IC50 of 1 48 nM 7 15 nM and 2 49 nM respectively Epertinib hydrochloride (S-222611 hydrochloride) exhibits antitumor activity