Learn More
Medchemexpress LLC Epertinib hydrochloride | 2071195-74-7 | >98.0% | 596.48 g/mol | C30H28Cl2FN5O3 | 1 ML
SDP
Catalog No. 5000241473
Encompass_Preferred
Epertinib hydrochloride is the hydrochloride salt of epertinib, a reversible and selective tyrosine kinase inhibitor of EGFR, HER2, and HER4 used in oncology research. The product is supplied as a 10 mM solution in DMSO (1 mL) and as solid powders; consult the datasheet and safety information for handling and purity details.
  • Potent, reversible inhibitor of EGFR, HER2, and HER4.
  • Suitable for in vitro and in vivo oncology research.
  • Available as a 10 mM DMSO solution for ready-to-use assays.
  • Also available as solid for formulation or long-term storage.
  • Molecular weight 596.48 g/mol and CAS number 2071195-74-7.
Catalog No. 50-002-41473 Supplier Medchemexpress LLC Supplier No. HY107367A10MM 1ML
Epertinib hydrochloride is the hydrochloride salt of epertinib, a reversible and selective tyrosine kinase inhibitor of EGFR, HER2, and HER4 used in oncology research. The product is supplied as a 10 mM solution in DMSO (1 mL) and as solid powders; consult the datasheet and safety information for handling and purity details.
  • Potent, reversible inhibitor of EGFR, HER2, and HER4.
  • Suitable for in vitro and in vivo oncology research.
  • Available as a 10 mM DMSO solution for ready-to-use assays.
  • Also available as solid for formulation or long-term storage.
  • Molecular weight 596.48 g/mol and CAS number 2071195-74-7.