Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Epertinib hydrochloride | 2071195-74-7 | 98.2% | 596.48 g/mol | C30H28Cl2FN5O3 | 100 MG
SDP

Catalog No. 5000239726
Encompass_Preferred

Epertinib hydrochloride is the hydrochloride salt of epertinib, a potent, orally active, reversible, and selective inhibitor of EGFR, HER2, and HER4. It is provided for preclinical research to evaluate signaling pathways and antitumor efficacy and is compatible with click-chemistry applications because of an alkyne functional group.

  • Potent, selective tyrosine kinase inhibitor of EGFR, HER2, and HER4.
  • Used in preclinical antitumor and signaling research.
  • Contains an alkyne moiety compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Supplied as a light yellow to yellow solid with high purity.
  • Stable under recommended storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).
  • Molecular weight 596.48 g/mol; molecular formula C30H28Cl2FN5O3.

Catalog No. 50-002-39726 Supplier Medchemexpress LLC Supplier No. HY107367A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Epertinib hydrochloride is the hydrochloride salt of epertinib, a potent, orally active, reversible, and selective inhibitor of EGFR, HER2, and HER4. It is provided for preclinical research to evaluate signaling pathways and antitumor efficacy and is compatible with click-chemistry applications because of an alkyne functional group.

  • Potent, selective tyrosine kinase inhibitor of EGFR, HER2, and HER4.
  • Used in preclinical antitumor and signaling research.
  • Contains an alkyne moiety compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Supplied as a light yellow to yellow solid with high purity.
  • Stable under recommended storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).
  • Molecular weight 596.48 g/mol; molecular formula C30H28Cl2FN5O3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.