Learn More
Medchemexpress LLC Epertinib hydrochloride | 2071195-74-7 | 98.2% | 596.48 g/mol | C30H28Cl2FN5O3 | 25 MG
SDP
Catalog No. 5000239694
Encompass_Preferred
Epertinib hydrochloride is a research-grade, orally active, reversible tyrosine kinase inhibitor selective for EGFR, HER2, and HER4. Supplied as a solid hydrochloride salt, it is intended for in vitro and preclinical research and should be handled according to its safety data sheet.
Catalog No. 50-002-39694 Supplier Medchemexpress LLC Supplier No. HY107367A25MG
Epertinib hydrochloride is a research-grade, orally active, reversible tyrosine kinase inhibitor selective for EGFR, HER2, and HER4. Supplied as a solid hydrochloride salt, it is intended for in vitro and preclinical research and should be handled according to its safety data sheet.