Learn More
Medchemexpress LLC Epertinib hydrochloride | 2071195-74-7 | MFCD32215297 | 98.2% | 596.48 g/mol | C30H28Cl2FN5O3 | 50 MG
SDP
Catalog No. 5000239946
Encompass_Preferred
Epertinib hydrochloride is a potent, orally active, reversible, and selective tyrosine kinase inhibitor of EGFR, HER2, and HER4 with low-nanomolar IC50 values and demonstrated antitumor activity in preclinical models. The hydrochloride salt is supplied as a solid research compound suitable for biochemical and cell-based studies and contains an alkyne moiety for click chemistry applications.
  • Potent inhibition of EGFR, HER2, and HER4 at low-nanomolar concentrations.
  • Demonstrated antitumor activity in preclinical in vivo models.
  • Contains an alkyne functional group for click chemistry conjugation.
  • Supplied as a solid research reagent with high purity.
  • Recommended sealed storage away from moisture to preserve stability.
Catalog No. 50-002-39946 Supplier Medchemexpress LLC Supplier No. HY107367A50MG
Epertinib hydrochloride is a potent, orally active, reversible, and selective tyrosine kinase inhibitor of EGFR, HER2, and HER4 with low-nanomolar IC50 values and demonstrated antitumor activity in preclinical models. The hydrochloride salt is supplied as a solid research compound suitable for biochemical and cell-based studies and contains an alkyne moiety for click chemistry applications.
  • Potent inhibition of EGFR, HER2, and HER4 at low-nanomolar concentrations.
  • Demonstrated antitumor activity in preclinical in vivo models.
  • Contains an alkyne functional group for click chemistry conjugation.
  • Supplied as a solid research reagent with high purity.
  • Recommended sealed storage away from moisture to preserve stability.