Learn More
Selleck Chemical LLC Epertinib hydrochloride
SDP
Supplier:  Selleck Chemical LLC S689725MG
Encompass_Preferred
Epertinib hydrochloride (S-222611 hydrochloride) is a potent orally active reversible and selective tyrosine kinase inhibitor of EGFR HER2 and HER4 with IC50 of 1 48 nM 7 15 nM and 2 49 nM respectively Epertinib hydrochloride (S-222611 hydrochloride) exhibits antitumor activity
Catalog No. 50-000-02483