Learn More
Medchemexpress LLC Epetraborole hydrochloride | 1234563-16-6 | 99.6% | 273.52 | C11H17BClNO4 | 10MG
SDP
Catalog No. 5000237105
Encompass_Preferred
Epetraborole hydrochloride is a research-grade leucyl-tRNA synthetase (LeuRS) inhibitor supplied as the hydrochloride salt for in vitro antibacterial research. It has a reported IC50 of 0.31 μM against LeuRS, is provided as a white to off-white solid with high purity (~99.6%), and is offered in small milligram pack sizes for early-stage studies.
  • Inhibits leucyl-tRNA synthetase with reported IC50 = 0.31 μM.
  • Supplied as the hydrochloride salt.
  • High purity appropriate for research use (approx. 99.6%).
  • White to off-white solid for straightforward handling and storage.
  • Available in milligram-scale pack sizes for screening and early research.
Catalog No. 50-002-37105 Supplier Medchemexpress LLC Supplier No. HY12479A10MG
Epetraborole hydrochloride is a research-grade leucyl-tRNA synthetase (LeuRS) inhibitor supplied as the hydrochloride salt for in vitro antibacterial research. It has a reported IC50 of 0.31 μM against LeuRS, is provided as a white to off-white solid with high purity (~99.6%), and is offered in small milligram pack sizes for early-stage studies.
  • Inhibits leucyl-tRNA synthetase with reported IC50 = 0.31 μM.
  • Supplied as the hydrochloride salt.
  • High purity appropriate for research use (approx. 99.6%).
  • White to off-white solid for straightforward handling and storage.
  • Available in milligram-scale pack sizes for screening and early research.