Learn More
Medchemexpress LLC Epetraborole hydrochloride | 1234563-16-6 | 99.6% | 273.52 | C11H17BClNO4 | 10MM 1ML
SDP
Catalog No. 5000239667
Encompass_Preferred
Epetraborole hydrochloride is a small-molecule leucyl-tRNA synthetase (LeuRS) inhibitor used in antibacterial research to block bacterial protein synthesis. Supplied as a concentrated solution (10 mM, 1 mL) for research use, it has formula C11H17BClNO4, molecular weight 273.52, and CAS 1234563-16-6.
  • Inhibits leucyl-tRNA synthetase, blocking protein synthesis.
  • Suitable for in vitro antibacterial assays and mechanism studies.
  • Provided as a concentrated solution for immediate use in assays.
  • High purity suitable for research applications.
  • Includes chemical identifiers for traceability and documentation.
Catalog No. 50-002-39667 Supplier Medchemexpress LLC Supplier No. HY12479A10MM 1ML
Epetraborole hydrochloride is a small-molecule leucyl-tRNA synthetase (LeuRS) inhibitor used in antibacterial research to block bacterial protein synthesis. Supplied as a concentrated solution (10 mM, 1 mL) for research use, it has formula C11H17BClNO4, molecular weight 273.52, and CAS 1234563-16-6.
  • Inhibits leucyl-tRNA synthetase, blocking protein synthesis.
  • Suitable for in vitro antibacterial assays and mechanism studies.
  • Provided as a concentrated solution for immediate use in assays.
  • High purity suitable for research applications.
  • Includes chemical identifiers for traceability and documentation.