Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Epetraborole hydrochloride | 1234563-16-6 | 99.6% | 273.52 g·mol⁻1 | C11H17BClNO4 | 50MG
SDP

Catalog No. 5000239234
Encompass_Preferred

Epetraborole hydrochloride is a boron-containing leucyl-tRNA synthetase inhibitor supplied for laboratory research into antibacterial mechanisms. The material is provided in small pack sizes for experimental use and is accompanied by batch COA and safety documentation for handling and quality verification.

  • Boron-containing leucyl-tRNA synthetase inhibitor used in antibacterial research.
  • Chemical name: Epetraborole hydrochloride.
  • CAS number: 1234563-16-6.
  • Molecular formula: C11H17BClNO4; molecular weight: 273.52 g·mol⁻1.
  • Purity (LCMS): 99.6%.
  • Reported bioactivity: leucyl-tRNA synthetase IC50 ≈ 0.31 μM.
  • Supplied in small research pack sizes, with batch COA and SDS available.

Catalog No. 50-002-39234 Supplier Medchemexpress LLC Supplier No. HY12479A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Epetraborole hydrochloride is a boron-containing leucyl-tRNA synthetase inhibitor supplied for laboratory research into antibacterial mechanisms. The material is provided in small pack sizes for experimental use and is accompanied by batch COA and safety documentation for handling and quality verification.

  • Boron-containing leucyl-tRNA synthetase inhibitor used in antibacterial research.
  • Chemical name: Epetraborole hydrochloride.
  • CAS number: 1234563-16-6.
  • Molecular formula: C11H17BClNO4; molecular weight: 273.52 g·mol⁻1.
  • Purity (LCMS): 99.6%.
  • Reported bioactivity: leucyl-tRNA synthetase IC50 ≈ 0.31 μM.
  • Supplied in small research pack sizes, with batch COA and SDS available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.