Learn More
Medchemexpress LLC Epetraborole hydrochloride | 1234563-16-6 | MFCD28411360 | 99.6% | 273.52 g/mol | C11H17BClNO4 | 100MG
SDP
Catalog No. 5000237104
Encompass_Preferred
Epetraborole hydrochloride is a research-grade leucyl-tRNA synthetase (LeuRS) inhibitor supplied as a hydrochloride salt for antibacterial research. It inhibits protein synthesis (reported IC50 = 0.31 μM) and is used in studies of multidrug-resistant gram-negative pathogens. Supplied as a high-purity solid, it requires sealed storage at 4°C and long-term storage in solution at -80°C.
Catalog No. 50-002-37104 Supplier Medchemexpress LLC Supplier No. HY12479A100MG
Epetraborole hydrochloride is a research-grade leucyl-tRNA synthetase (LeuRS) inhibitor supplied as a hydrochloride salt for antibacterial research. It inhibits protein synthesis (reported IC50 = 0.31 μM) and is used in studies of multidrug-resistant gram-negative pathogens. Supplied as a high-purity solid, it requires sealed storage at 4°C and long-term storage in solution at -80°C.