Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eph inhibitor 1 10mg | 861249-77-6 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY11419910MG

Encompass_Preferred

Eph inhibitor 1 is a small-molecule Ephrin (Eph) receptor inhibitor supplied for research use. It is a white to light yellow solid with high purity and DMSO solubility, intended for biochemical and cellular studies related to Eph signaling and neurological disorder research.

  • Potent Ephrin receptor inhibitor for research use.
  • High purity (99.92%).
  • Molecular weight 303.32 g/mol; formula C17H13N5O.
  • White to light yellow solid; DMSO solubility ~100 mg/mL; may require ultrasonication.
  • Available in small milligram pack sizes suitable for screening and assay development.
  • Store at 4°C protected from light; in solution store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-08159


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.