Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eph inhibitor 1 5mg | 861249-77-6 | 5 MG
SDP

Catalog No. 5000229844
Encompass_Preferred

Eph inhibitor 1 is a small-molecule Eph (ephrin) receptor inhibitor for research use. It is supplied as a high-purity solid and has potential applications in neurological disorder research.

  • Potent Eph receptor inhibitor.
  • Molecular formula C17H13N5O.
  • Molecular weight 303.32 g/mol.
  • Purity 99.92% (as reported by manufacturer).
  • Available in small pack sizes for laboratory use.
  • Store at 4°C and protect from light.

Catalog No. 50-002-29844 Supplier Medchemexpress LLC Supplier No. HY1141995MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eph inhibitor 1 is a small-molecule Eph (ephrin) receptor inhibitor for research use. It is supplied as a high-purity solid and has potential applications in neurological disorder research.

  • Potent Eph receptor inhibitor.
  • Molecular formula C17H13N5O.
  • Molecular weight 303.32 g/mol.
  • Purity 99.92% (as reported by manufacturer).
  • Available in small pack sizes for laboratory use.
  • Store at 4°C and protect from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.