Learn More
ATCC EpH4-Ev; Mammary Epithelium; Mouse (Mus musculus)
Catalog No. 502381780
Encompass
EpH4-Ev is an epithelial cell line isolated from the mammary glands of a mouse. It can be used to study normal epithelial cell function and cell transformation.
Catalog No. 50-238-1780 Supplier ATCC Supplier No. CRL3063
EpH4-Ev is an epithelial cell line isolated from the mammary glands of a mouse. It can be used to study normal epithelial cell function and cell transformation.