Learn More
Carnabio Usa Inc EPHA1/10UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 0811910
Encompass
EPHA1, Cytoplasmic domain, Wild type, Amino Acid 586-976, NP_005223.3, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0768