Learn More
Carnabio Usa Inc EPHA1/5UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 081195
Encompass
EPHA1, Cytoplasmic domain, Wild type, Amino Acid 586-976, NP_005223.3, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0413