Learn More
Enzo Life Sciences Human Ephrin Type A receptor 2 Recombinant monoclonal antibody (1C1) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5420200
Encompass_Preferred
Clone: 1C1. Isotype: IgGκ. Immunogen: 1C1 was isolated after screening of a non-immunized human Fab phage library. Selection was based on the ability to bind soluble human recombinant EphA2 (residues 183-510). The Fab was subsequently converted to full length human IgG1. Species reactivity: Human. Specificity: Recognizes the N-termianal LBD (ligand-binding domain) region of the EphA2 ectodomain (IgG1 KD ~0.8 nM; Fab KD ~140 nM) and not to other related human Eph receptors such as EphA1, EphA2, and EphA4, or any EphB receptor family members. Applications: ELISA, Flow Cytometry, IF, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3068