Learn More
Medchemexpress LLC EphA2 Human Biotin 100ug
SDP
Catalog No. 5000180759
Encompass_Preferred
The EphA2 protein is an important member of the ephrin receptor subfamily that mediates developmental events particularly in the nervous system It has a single kinase domain and an extracellular region that binds ephrin-A ligands EphA2 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived EphA2 protein expressed by HEK293 with C-6 His C-Avi labeled tag
Catalog No. 50-001-80759 Supplier Medchemexpress LLC Supplier No. HYP72992100UG
The EphA2 protein is an important member of the ephrin receptor subfamily that mediates developmental events particularly in the nervous system It has a single kinase domain and an extracellular region that binds ephrin-A ligands EphA2 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived EphA2 protein expressed by HEK293 with C-6 His C-Avi labeled tag