Learn More
Sino Biological Recombinant Human EphA2 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 13926H27HB20
Encompass_Preferred
A DNA sequence encoding the human EPHA2 (NP_004422.2) (Met1-Asn534) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5206