Learn More
Eurofins Discoverx EphA3, active, 250 µg. (14-644M)
Supplier:  Eurofins Discoverx 14644M
Encompass
N-terminal 6xHis-tagged recombinant, human EphA3 amino acids 578-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9110