Learn More
Sino Biological Recombinant Mouse EphA1 Protein (GST Tag), Active
Catalog No. 502575107
Encompass_Preferred
Recombinant mouse EPHA1 (569-end) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
Catalog No. 50-257-5107 Supplier Sino Biological Supplier No. E1311G50
Recombinant mouse EPHA1 (569-end) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.