Learn More
Aviva Systems Biology Corp Epha4 Antibody - C-terminal region (ARP61663_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61663_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Epha4. It was validated on Western Blot. . Alternate names: 2900005C20Rik, AI385584, Cek8, Hek8, Sek, Sek1, Tyro1, rbApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-434