Learn More
ACROBiosystems ACROBiosystems Human EphA4 Protein, His Tag
Catalog No. 501956492
Encompass_Preferred
Recombinant Protein;100UG;Human EphA4, His Tag (EP4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Thr 547 (Accession # P54764-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.3 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human EphA4, His Tag (Cat. No. EP4-H52H8) at 5 μg/mL (100 μL/well) can bind Human Ephrin-A4, Fc Tag (Cat. No. EA4-H5250) with a linear range of 0.6-5 ng/mL (QC tested).
Catalog No. 50-195-6492 Supplier ACROBiosystems Supplier No. EP4H52H8100UG
Recombinant Protein;100UG;Human EphA4, His Tag (EP4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Thr 547 (Accession # P54764-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.3 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human EphA4, His Tag (Cat. No. EP4-H52H8) at 5 μg/mL (100 μL/well) can bind Human Ephrin-A4, Fc Tag (Cat. No. EA4-H5250) with a linear range of 0.6-5 ng/mL (QC tested).