Learn More
ECM Bioscience EphA4 (Tyr-602))[conserved site], phospho-specific Rabbit Polyclonal Antibody, ECM Biosciences
Catalog No. 501720776
Encompass_Preferred
This antibody was cross-adsorbed to unrelated phosphotyrosine peptides before affinity purification using EphA4 (Tyr-602) peptide. The purified antibody detects a 120 kDa* band corresponding to EphA4 in Western blots of HUVEC cells treated with pervanadate and in mouse brain tissue. The immunogen was EphA4 (Tyr-602) synthetic peptide (coupled to carrier protein) corresponds to amino acids surrounding phosphorylated tyrosine 602 in human EphA4. This sequence has significant homology to the conserved site in rat and mouse EphA4, and is conserved well in most other EphA and EphB family members. Accession #P54764. Applications = WB, E, IHC; Species Reactivity = Hu, Rt, Ms, Ck; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = SEK, Eph
Catalog No. 50-172-0776 Supplier ECM Bioscience Supplier No. EP2731
Add to Cart
Add to Cart
This antibody was cross-adsorbed to unrelated phosphotyrosine peptides before affinity purification using EphA4 (Tyr-602) peptide. The purified antibody detects a 120 kDa* band corresponding to EphA4 in Western blots of HUVEC cells treated with pervanadate and in mouse brain tissue. The immunogen was EphA4 (Tyr-602) synthetic peptide (coupled to carrier protein) corresponds to amino acids surrounding phosphorylated tyrosine 602 in human EphA4. This sequence has significant homology to the conserved site in rat and mouse EphA4, and is conserved well in most other EphA and EphB family members. Accession #P54764. Applications = WB, E, IHC; Species Reactivity = Hu, Rt, Ms, Ck; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = SEK, Eph