Learn More
Sino Biological Recombinant Human EPHA5 Protein (GST Tag), Active
Catalog No. 5000083645
Encompass_Preferred
Recombinant human EPHA5 (662-948) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-000-83645 Supplier Sino Biological Supplier No. E1711G10
Recombinant human EPHA5 (662-948) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.