Learn More
Eurofins Discoverx EphA5, active, 250 µg. (14-639M)
Supplier:  Eurofins Discoverx 14639M
Encompass
N-terminal 6xHis-tagged recombinant, human EphA5 amino acids 655-956. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9100