Learn More
Eurofins Discoverx EphA7, active, 250 µg. (14-672M)
Supplier:  Eurofins Discoverx 14672M
Encompass
N-terminal 6xHis-tagged recombinant, human EphA7 amino acids 613-909. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9162