Learn More
Carnabio Usa Inc EPHB1/10UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 0812810
Encompass
EPHB1, Cytoplasmic domain, Wild type, Amino Acid 578-984, NP_004432.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0776