Learn More
Carnabio Usa Inc EPHB2/5UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 081295
Encompass
EPHB2, Cytoplasmic domain, Wild type, Amino Acid 581-987, NP_004433.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0422