Learn More
Sino Biological Recombinant Human EphA4 Protein (GST Tag), Active
Catalog No. 502575094
Encompass_Preferred
Recombinant human EPHA4 (610-887) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5094 Supplier Sino Biological Supplier No. E1611G500
Recombinant human EPHA4 (610-887) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.