Learn More
Eurofins Discoverx EphB3, active, 10 µg. (14-561)
Supplier:  Eurofins Discoverx 14561
Encompass
N-terminal 6xHis-tagged recombinant, human EphB3 amino acids 599-920. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8973