Learn More
Aviva Systems Biology Corp EPHB3 Antibody - C-terminal region (ARP63815_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63815_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EPHB3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human EPHB3. Alternate names: ETK2, HEK2, TYRO6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-093