Learn More
Medchemexpress LLC EphB4 Human HEK293 10ug
SDP
Catalog No. 5000179061
Encompass_Preferred
EphB4 protein is a receptor tyrosine kinase that binds to ephrin B ligand and initiates bidirectional signaling Positive signaling regulates the rejection and separation of cells from EFNB2-expressing cells EphB4 Protein Human (HEK293 Fc) is the recombinant human-derived EphB4 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-79061 Supplier Medchemexpress LLC Supplier No. HYP7417110UG
EphB4 protein is a receptor tyrosine kinase that binds to ephrin B ligand and initiates bidirectional signaling Positive signaling regulates the rejection and separation of cells from EFNB2-expressing cells EphB4 Protein Human (HEK293 Fc) is the recombinant human-derived EphB4 protein expressed by HEK293 with C-hFc labeled tag