Learn More
Medchemexpress LLC EphB4 Mouse HEK293 10ug
Supplier: Medchemexpress LLC HYP7300510UG
Ephb4 Protein a receptor tyrosine kinase plays a critical role in angiogenesis and development Dysregulation of Ephb4 Protein has been associated with disorders like cancer and cardiovascular diseases Targeting Ephb4 Protein may provide potential therapeutic interventions in these conditions by modulating angiogenesis inhibiting tumor growth and managing cardiovascular complications EphB4 Protein Mouse (HEK293 His) is the recombinant mouse-derived EphB4 protein expressed by HEK293 with C-His labeled tag