Learn More
ACROBiosystems Recombinant Protein;Human EphB4 Protein;HEK293;1MG;EP4,EphB4,HTK,MYK1,TYRO11,EphB-4,MYK-1
Catalog No. 501657565
Encompass_Preferred
Recombinant Protein;Human EphB4 Protein;1MG;Human EphB4, His Tag (EP4-H5229) is expressed from human 293 cells (HEK293). It contains AA Glu 17 - Ala 539 (Accession # NP_004435.3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.8 kDa. The protein migrates as 65-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7565 Supplier ACROBiosystems Supplier No. EP4H52291MG
Recombinant Protein;Human EphB4 Protein;1MG;Human EphB4, His Tag (EP4-H5229) is expressed from human 293 cells (HEK293). It contains AA Glu 17 - Ala 539 (Accession # NP_004435.3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.8 kDa. The protein migrates as 65-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.