Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human EphB4, ultra sensitivity (primary amine labeling);HEK293;200UG;EP4,EphB4,HTK,MYK1,TYRO11,EphB-4,MYK-1
Supplier:  ACROBiosystems EP4H8229200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human EphB4, ultra sensitivity (primary amine labeling);200UG;MABSol Biotinylated Human EphB4 (EP4-H8229) is expressed from human HEK293 cells. It contains AA Leu 16 - Ala 539 (Accession # NP_004435.3). It is the biotinylated form of Human EphB4 protein, His Tag (Cat # EP4-H5229).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.8 kDa. The protein migrates as 65-70 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.
Catalog No. 50-165-9176