Learn More
ACROBiosystems ACROBiosystems Human Ephrin-A4 / EFNA4 Protein, His Tag
Catalog No. 501957472
Encompass_Preferred
Recombinant Protein;1MG;Human Ephrin-A4, His Tag (EA4-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession # AAI07484).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.5 kDa. The protein migrates as 20-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7472 Supplier ACROBiosystems Supplier No. EA4H52291MG
Recombinant Protein;1MG;Human Ephrin-A4, His Tag (EA4-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession # AAI07484).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.5 kDa. The protein migrates as 20-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.