Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ephrin-B1 (EFNB1) recombinant human C-terminal Fc fusion protein | 97.1% | 500UG
SDP

Catalog No. 5000177886
Encompass_Preferred

Recombinant human Ephrin-B1 extracellular domain expressed in HEK293 cells with a C-terminal human Fc tag, supplied lyophilized for research use. Intended for receptor-ligand binding assays, signaling studies, ELISA, and cell-based assays.

  • Expression host: HEK293.
  • Tag: C-terminal human Fc (C-hFc).
  • Supplied form: lyophilized powder.
  • Available sizes: 10 μg, 50 μg, 100 μg, 500 μg, 1 mg.
  • Purity: 97.1% as reported by distributor.
  • Molecular weight: ~58.8 kDa (Fc fusion).
  • Applications: receptor binding, signaling studies, ELISA, cell stimulation.
  • Storage recommendation: aliquot and store at -20°C or -80°C for long term.

Catalog No. 50-001-75923 Supplier Medchemexpress LLC Supplier No. HYP700609500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Ephrin-B1 extracellular domain expressed in HEK293 cells with a C-terminal human Fc tag, supplied lyophilized for research use. Intended for receptor-ligand binding assays, signaling studies, ELISA, and cell-based assays.

  • Expression host: HEK293.
  • Tag: C-terminal human Fc (C-hFc).
  • Supplied form: lyophilized powder.
  • Available sizes: 10 μg, 50 μg, 100 μg, 500 μg, 1 mg.
  • Purity: 97.1% as reported by distributor.
  • Molecular weight: ~58.8 kDa (Fc fusion).
  • Applications: receptor binding, signaling studies, ELISA, cell stimulation.
  • Storage recommendation: aliquot and store at -20°C or -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.