Learn More
Medchemexpress LLC Ephrin-B1/EFNB1 Rat 100ug
SDP
Catalog No. 5000180469
Encompass_Preferred
Ephrin-B1/EFNB1 proteins bind to Eph receptors on neighboring cells and promote bidirectional signaling in developing neurons blood vessels and epithelia It has high affinity for EPHB1/ELK also binds EPHB2 and EPHB3 and induces commissural axon/growth cone collapse Ephrin-B1/EFNB1 Protein Rat (HEK293 His) is the recombinant rat-derived Ephrin-B1/EFNB1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80469 Supplier Medchemexpress LLC Supplier No. HYP73030100UG
Ephrin-B1/EFNB1 proteins bind to Eph receptors on neighboring cells and promote bidirectional signaling in developing neurons blood vessels and epithelia It has high affinity for EPHB1/ELK also binds EPHB2 and EPHB3 and induces commissural axon/growth cone collapse Ephrin-B1/EFNB1 Protein Rat (HEK293 His) is the recombinant rat-derived Ephrin-B1/EFNB1 protein expressed by HEK293 with C-His labeled tag