Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ephrin-B2 (EFNB2) protein, canine | >95.0% | 100 UG
SDP

Catalog No. 5000292841
Encompass_Preferred

Recombinant canine Ephrin-B2 (EFNB2) extracellular domain expressed in HEK293 cells and provided as a lyophilized His-tagged protein for research applications including receptor binding and signaling studies.

  • Produced in HEK293 mammalian expression system.
  • C-terminal His tag for detection and purification.
  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~25-35 kDa due to glycosylation.
  • Lyophilized formulation with recommended aliquoting and storage at -20°C or -80°C for long term.
  • Sequence corresponds to residues 128-229 of the canine EFNB2 extracellular region.

Catalog No. 50-002-92841 Supplier Medchemexpress LLC Supplier No. HYP75230100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant canine Ephrin-B2 (EFNB2) extracellular domain expressed in HEK293 cells and provided as a lyophilized His-tagged protein for research applications including receptor binding and signaling studies.

  • Produced in HEK293 mammalian expression system.
  • C-terminal His tag for detection and purification.
  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~25-35 kDa due to glycosylation.
  • Lyophilized formulation with recommended aliquoting and storage at -20°C or -80°C for long term.
  • Sequence corresponds to residues 128-229 of the canine EFNB2 extracellular region.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.