Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ephrin Receptor B1 Antibody (YA2176) | 50 UL
SDP

Catalog No. 5000444430
Encompass_Preferred

This Ephrin Receptor B1 Antibody (YA2176) is a non-conjugated liquid antibody, produced in rabbit. It is suitable for laboratory applications, including Western Blot (WB) and Immunoprecipitation (IP), and reacts with human, mouse, and rat samples. It is formulated for stability and comes with specific dilution recommendations for various uses.

  • Produced in rabbit
  • Reacts with human, mouse, rat
  • Non-conjugated
  • Applications include Western Blot and Immunoprecipitation
  • Recommended dilution ratio for WB is 1:500-1:1000
  • Recommended dilution ratio for IP is 1:50
  • Supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Store at -20°C for 1 year
  • Avoid repeated freeze / thaw cycles
  • Shipped with blue ice
  • Predicted molecular weight of 110 kDa; observed molecular weight of 120 kDa

Catalog No. 50-004-44430 Supplier Medchemexpress LLC Supplier No. HYP8243150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Ephrin Receptor B1 Antibody (YA2176) is a non-conjugated liquid antibody, produced in rabbit. It is suitable for laboratory applications, including Western Blot (WB) and Immunoprecipitation (IP), and reacts with human, mouse, and rat samples. It is formulated for stability and comes with specific dilution recommendations for various uses.

  • Produced in rabbit
  • Reacts with human, mouse, rat
  • Non-conjugated
  • Applications include Western Blot and Immunoprecipitation
  • Recommended dilution ratio for WB is 1:500-1:1000
  • Recommended dilution ratio for IP is 1:50
  • Supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Store at -20°C for 1 year
  • Avoid repeated freeze / thaw cycles
  • Shipped with blue ice
  • Predicted molecular weight of 110 kDa; observed molecular weight of 120 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.