Learn More
Eurofins Discoverx EphrinA3, 200 µg. (92-1142)
Supplier:  Eurofins Discoverx 921142
Encompass
Ephrin-A3 is a membrane protein tyrosine kinase and member of the ephrin (EPH) family. Implicated in cell migration, repulsion, and adhesion during neuronal, vascular, and epithelial development. Potent agonist of the EPH receptors. Provided as 200 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9674