Learn More
ACROBiosystems Recombinant Protein;Human Ephrin-A3 / EFNA3 Protein, Fc Tag;HEK293;1MG;EA3,Ephrin-A3,EFL-2,EHK1-L,LERK-3,EphrinA3,EFL2,EPLG3,LERK3,EFNA3
Catalog No. 501658762
Encompass_Preferred
Recombinant Protein;Human Ephrin-A3 / EFNA3 Protein, Fc Tag;1MG;Human Ephrin-A3, Fc Tag (EA3-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 213 (Accession # AAI10407).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8762 Supplier ACROBiosystems Supplier No. EA3H52581MG
Recombinant Protein;Human Ephrin-A3 / EFNA3 Protein, Fc Tag;1MG;Human Ephrin-A3, Fc Tag (EA3-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 213 (Accession # AAI10407).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.